Normalized Detection Fractions (NDF)
Normalized Detection Fractions provide the color-coding of calls in heatmaps.
Normalized Detection Fractions are interpreted as follows:
-
Higher values indicate a greater likelihood that the fusion call is correct and that the fusion is present in your sample.
-
Lower values indicate a greater likelihood that the fusion is not present in your sample.