Locus View versus Allele View of variants
When you set workflow parameters, you can set the view that is used for variants in analysis results with the Variant View parameter. You have the option to view variants per position, Locus View, or variants per allele, Allele View. For more information on how to set this view, see Customize tuning parameters and Variant Finding parameters. The VCF files for both the Locus View and Allele View include one line for each variant row that is displayed in the Ion Reporter™ Software Analysis Results table.
The FDP (Flow Evaluator read depth at the locus) and DP (Total read depth at the locus) are subfields inside the INFO field of the VCF file. The values of these two fields remain the same for both the Locus View VCF file and the Allele View VCF file. These two fields, FDP and DP, are based on the TOTAL SUM of ALL the FAO (Flow Evaluator Alternate allele observations) and AO (Alternate allele observations) fields, which are based on the following formulas:
FDP=SUM(FAO)+FRO or DP=SUM(AO)+RO