You can hide or unhide variants that are
listed in analysis results in Ion Reporter™ Software. It is useful to hide variants if
you do not want to include them in visualizations of the analysis results, in downloads
of the results, or in reports that you generate from the results.
Click
the Analyses tab to open the list of analyses, then click an
analysis of interest.
The Analysis Results table opens.
To hide variants, select the variants that you want to hide.
Click Selected Variants
> Hide.
The selected variants are removed from the
Analysis Results screen and are listed in the count of
Hidden Variants in the Filter Options
pane.
To unhide variants, click Hidden Variants in the
Filter Options pane, then click Selected Variants
> Unhide.
Hidden variants are not visible on the Analysis Results, and are
not included in the results files when you download variants, or in visualized results.