Download an amplicon coverage report
You can download an amplicon coverage report in Ion Reporter™ Software. Amplicon coverage reports contain details such as amplicon coverage, chromosome location, and start and end positions.
  1. In the Analysis tab, click Overview.
  2. Select the row of the analysis that you want to report on.
  3. Click (Actions)View Coverage Report.
  4. Click Export to generate a tab-separated CSV file that contains details such as chromosome location, and start and end positions on forward and reverse strands.
    The downloaded file is named ampliconmetrics.csv.
Amplicon coverage report metrics
The amplicon coverage report includes the following metrics:
Metric
Description
Region
The amplicon name as defined in the BED file for the amplicon region.
Chr
Chromosome
Start
The start coordinate of the amplicon.
End
the end coordinate of the amplicon.
Attributes
The attribute in the BED file for the amplicon.
TotalE2E
The number of assigned reads from end to end for the amplicon region.
FwdE2E
The number of assigned forward strand reads from end to end for the amplicon region.
RevE2E
The number of assigned reverse strand reads from end to end for the amplicon region.
Total
The total number of reads assigned to this amplicon. This value is equal to Fwd + Rev.
Fwd
The number of forward strand reads assigned to this amplicon. This value may differ from the reads for end to end.
Rev
The number of reverse strand reads assigned to this amplicon. This value may differ from the reads for end to end.