This filter can be added to filter chains in the software to detect known variants in the Ion AmpliSeq™ Community Panel for the cystic fibrosis transmembrane regulator (CFTR) gene, for example: c.2054delA.
Filter conditions include the ability to select all named variants, to select specific named variants, and to include or exclude unannotated variants in filtered analysis results.
The filter returns variants in samples that match the CFTR variants that are specified in the filter.