DrugBank filter
This filter can be added to filter chains in the software to detect variants that are correlated with drugs and drug targets listed DrugBank database. The specific filters are:
Filter conditions include the ability to select specific values, to include all DrugBank values (In), or to exclude all DrugBank values (Not In). In addition, unannotated variants can be included or excluded in filtered analysis results.
This filter detects variants that are correlated with drugs and drug targets in DrugBank, which can be included in or excluded from filtered analysis results.
×