UCSC Common SNPs filter
This filter can be added to filter chains in the software to detect variants that match those in the UCSC Genome Browser Common SNPs database.
Filter conditions include the ability to include (In) or exclude (Not In) variants that match the UCSC Common SNPs in filtered results.
The filter returns variants in samples that match the UCSC Common SNPs database.
×