Analysis workflows for Oncomine™ assays and analysis workflow templates for Ion AmpliSeq™ HD panels in Ion Reporter™ Software include the Oncomine™ Variant Annotator plugin. The plugin integrates into analysis results data from more than 24,000 exomes across solid tumor and hematological cancer types, and annotates variants relevant to cancer with Oncomine™ Gene class and Oncomine™ Variant class information. For more information and a full list of annotation rules for each Oncomine™ assay, see Oncomine Variant Annotator plugin criteria, or contact your local support representative, Field Bioinformatics Specialist (FBS), or Clinical Account Consultant (CAC).
Use the Oncomine™ Variant Annotator plugin with Oncomine™ panels only.