Change Y-axis height in Whole Genome aneuploidy visualization
You can expand the height of the Y-axis when you view whole genome aneuploidy analysis results in Ion Reporter™ Genomic Viewer (IRGV). By default the image of the data is 300 pixels high. IRGV includes the option to double this height to 600 pixels. The expanded view is useful when you review mosaic data, because more data is shown on a single plot and more space is available between data points, making the mosaic data more pronounced.
  1. Under the Analyses tab, click Overview.
  2. Select one or more analyses in the Analysis Results screen, then click Visualize.
  3. In the IRGV & Generate Report tab, scroll to the bottom of the visualization, then click IRGV/Export & PreferencesShow IRGV preferences to expand the Preferences section.
    Option
    Description
    Expanded
    Select this option to increase the height of the Whole Genome view to 600 pixels.
    Compressed
    This is the default setting. Select this option to set the height of the Whole Genome view to 300 pixels.