JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Delete an analysis note You can delete an analysis note in Ion Reporter™ Software. Analysis notes record any written comments or observations for an analysis that has successfully completed. In the Analyses tab, select Overview. In the row of an analysis of interest, click (Note). In the Analysis Notes dialog box, select the note that you want to delete, then click (Delete). Click Done. Related Topics Parent topic: Add an analysis note ×