There are two ways to monitor analyses in Ion Reporter™ Software.
In the Analyses tab, click Overview, then review the Status column.
The analysis is reported as Pending, % complete, Successful , or Failed.
You can also check for email notifications. Ion Reporter™ Software sends an email notification when the analysis is complete, and indicates whether the analysis was successful or failed.