Genetic Category Type filter
This filter can be included in a filter chain in the software to detect variants of specific genetic category types.
Filter conditions include the ability to select all, none, or specific genetic category type annotations, and to include or exclude unannotated variants in filtered analysis results.
Genetic category types include:
-
HasDeNovoNonRefAllele
-
HasDeNovoRefAllele
-
IsNewlyHomzygousNonRef
-
IsNewlyHomozygousRef
-
InCompoundHeterozygote
-
InTransPhaseCompoundHeterozygote
-
HasMaleMaternalX
-
HasUnknownX
-
InconsistentWithFather
-
InconsistentWithMother
-
InconsistentWithParents
This filter returns genetic category type annotations for the variants included in the samples.