This filter can be used in a filter chain in the software to detect INDELs of specific homopolymer lengths.
Filter conditions include the ability to set a range from 0 to 1,000 for homopolymer length. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
This filter returns INDEL variants in samples that meet the specified homopolymer length.