View the Mutational Signature Prediction report
To view the mutational signature prediction report (signature_identification_<analysis_name>_report.pdf) you must first download the ZIP folder. For more information, see Download filtered variants. The folder contains the following files.
PDF Report
Profile of Mutational signature of Sample (Only available if a mutational signature match is found.)
Profile of enriched COSMIC signature (One or More, only available if a mutational signature match is found.)
deconstructSigs Weights Pie Chart (Only available if a mutational signature match is found.)
Trinucleotide distribution of Variants
×