Get the latest updates from the MyVariants database
You can get the latest additions, deletions, or changes to the MyVariants database for an organization by synchronizing the analysis results to the MyVariants database in Ion Reporter™ Software. For example, you can open older analysis results and apply the updates for any MyVariants that were added, deleted, or changed, to the older analysis results.
  1. In the Analyses tab, click Overview.
  2. Click the link of a new or existing analysis to open the Analysis Results.
  3. Click  MyVariants to download flags from the MyVariants database into the analysis results.
  4. (Optional) Manually edit Important or Ignore flags within the analysis results.