Example
RNA Exon Tile Fusion Imbalance
read count plot for a single analysis
RNA Exon Tile Fusion Imbalance
plots provide a visual representation of the RNA fusion imbalance analysis results
for the detection of novel fusions in a partner agnostic manner. The
Normalized Read Depth by Gene
plot shows the mean read counts of each gene that is captured on the chip for the
selected sample. For each gene, the read counts are normalized to the number of
amplicons.