JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. View 3'/5' Imbalance scores You can detect novel fusions of select driver genes in Ion Reporter™ Software with 3'/5' Imbalance scores that are shown in analysis results. In the Analyses tab, click Overview. Click an analysis name. The Analysis Results screen opens to the Fusions table. The 3'/5' imbalance values are listed in the 3'/5' Imbalance column of the Fusions table. Related Topics Parent topic: 3'/5' Imbalance scores for assay calls ×