Example Normalized Read Depth by GeneRNA Exon Tile Fusion Imbalance
plot for multiple analyses
RNA Exon Tile Fusion Imbalance
plots provide a visual representation of the RNA fusion imbalance analysis results
for the detection of novel fusions in a partner agnostic manner. The
Normalized Read Depth by Gene
plot shows the mean read counts of each gene that is captured on the chip. For each
gene, the read counts are normalized to the number of amplicons. For multi-analysis
visualization, the data are displayed side by side. The number of bars corresponds
to the number of analyses.