HasUnknownX
A proband variant is assigned the category HasUnknownX if all of the following are true:
-
The proband is male.
-
The variant is on the X chromosome.
-
The variant is hemizygous (non-reference).
-
The father's genotype at this variant's position is hemizygous reference.
-
The mother's genotype does not include the proband variant's allele.
-
The variant's functional annotations include at least one in unknown_x_functional_types.
-
The frequency of the proband's minor allele is less than unknown_x_max_population_allele_frequency.
×