Immune repertoire analysis results report the CDR3 lengths of clones that are identified in a
sample. Select
CDR3 histogram
from the Views
list to see a histogram that represents the data.
Relative frequency (Y‑axis) of identified clones with a given CDR3 nucleotide length (X‑axis)
The histogram indicates the distribution of CDR3 lengths for clones that are identified in the sample.