Open and review analysis results
- In the Analysis tab, click Overview.
- In the Analyses table, click the name of the analysis for which you want to view analysis results.
Analyses that are available to view have an active link. You cannot view analyses that are still running.
The Analysis Results table opens to display a list of variants and detailed information about the variants. You can customize the view of the analysis results. For more information, see Customize the Analysis Results table Edit Analysis Results.
The Allele Coverage column reports counts for all reported alleles, not only genotype alleles. In contrast, the Allele Read Count filter sets the minimum count of the genotype alleles.
- Click one of the following views above the list of variants to display a different view of variants, annotations and other analysis results data: For more information about the data contained in the Analysis Results table, see Detailed analysis metrics.
×