Add a note to a variant
- In the Analysis tab, click Overview.
- Click an analysis name to open the Analysis Results screen.
-
In the Analysis Results screen, click
or
in the Variant Details column (second from the leftmost column).
-
In the Variant Details screen, click Notes.
A appears.
- In a new Variant Details screen, in Note text, enter the note.
- Click Add Note.
×