Immune repertoire analysis results
Analyses that are performed in Ion Reporter™ Software with Oncomine™ immune repertoire analysis workflows report the frequency and sequence information for clonotypes, and provide other data results, such as clone frequency and Jaccard Similarity indices. There are interactive graphs and plots that you can use to adjust views of the data and access details about clones in the software. You can also download the graphs and plots that you have adjusted, or other static graphics that are available in PNG or PDF file formats. You can download static plots in PDF format that can be added to slides for presentations.
The immune repertoire analysis results also include sample and quality control results for each sample that is included in the analysis.