View immune repertoire analysis results
If you use an immune repertoire analysis workflow, you can view the analysis results in Ion Reporter™ Software.
To perform a multi-sample analysis, see Compare the immune repertoire between samples.
  1. In the Analyses tab, click Overview.
  2. In the Workflow list, select an immune repertoire analysis workflow to limit the list of results to immune repertoire analyses.
    You can further refine the list of analyses with other filters, or click column headings to sort the list.
  3. Click the analyses name hyperlink.
  4. In the Immune Repertoire Results screen, click the hyperlinked Sample name to open the Immune Repertoire Results for that sample.
  5. Select the Sample Results, Sample QC, or Plugin Results tab, then select the graphical representation of the data from the Views list.