Delete custom annotation sources
An administrator can view and delete custom annotation sources that were created by users. Custom annotation sets created by users are displayed in the Workflow Presets screen.
Predefined annotation sources cannot be deleted. If a custom annotation source is used in an analysis workflow, it cannot be deleted.
- In the Workflows tab, click Presets.
-
From the presets list, click
Annotation Sets,
then select
Annotation Source.
A table opens that lists the custom annotation sources. It shows information about the annotation, such as: name, version, source version, reference, created by, created on, and source type.
-
In the Workflow Presets list, click the custom annotation source that you want to delete, then click .
The annotation source is removed from the Workflow Presets list.
)