The Total Mapped Fusion Panel Reads is a QC metric that is reported for gene fusion analyses.
This metric reports the sum of all reads that are valid and are assigned to any assay in
the panel. Unmapped reads and poorly aligned reads are excluded from this total.