Customize tuning parameters
You can modify tuning parameters in the Parameters workflow step during analysis workflow creation in Ion Reporter™ Software.
All analysis workflow types allow some parameter user-defined settings. The parameters that you can customize vary by analysis workflow. Parameter categories include: annotation, bamstats, CNV finding, MSI, read mapping, and variant finding.
In the Parameters workflow step, select the parameter category on the left, then confirm the settings or change the settings if needed.