You can add sample attributes to the Samples table that you view in Ion Reporter™ Software.
In the Samples tab, click Overview.
Click Preferences
> Select Columns, then select the sample attributes that you want to add to the
Samples table,
or deselect the sample attributes that you want to remove from the table.
Custom attributes are denoted with an asterisk
(*) in the
available columns list.
The selected sample attributes are added to the Samples table until you close the table. If you create a sample CSV file before you close the table, each custom attribute is included in the CSV file.