Oncomine™ Colon cfDNA Assay
Variant type |
Annotation criteria |
||
|---|---|---|---|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
Loss-of-function truncating hotspot mutation |
Loss-of-Function |
Truncating |
|
ERBB2 exon 20 insertion[1] |
Gain-of-Function |
ERBB2Exon20Insertion |
1 For more information, see Ion Torrent Class-Based Variants
×