Oncomine™ Myeloid Assay
Variant type |
Annotation criteria |
||
|---|---|---|---|
Gene fusion |
Gain-of-Function |
Fusion |
|
Non-targeted fusion |
Gain-of-Function |
NonTargetedFusion |
|
RNA exon variant |
Gain-of-Function |
RNAExonVariant |
|
Loss-of-function truncating de novo mutation |
Loss-of-Function |
Truncating |
|
Missense hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
In-frame hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
Promoter hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
Gain-of-function truncating hotspot mutation |
Gain-of-Function |
Hotspot |
|
Loss-of-function truncating hotspot mutation |
Loss-of-Function |
Truncating |
|
MNV hotspot mutation |
Gain-of-Function Loss-of-Function |
Hotspot |
|
JAK2 exon 12 indel[1] |
Gain-of-Function |
JAK2Exon12Indel |
|
KIT exon 11 deletion1 |
Gain-of-Function |
KITExon11Deletion |
|
KIT exon 11 insertion1 |
Gain-of-Function |
KITExon11Insertion |
|
FLT3 internal tandem duplication |
Gain-of-Function |
FLT3ITD |