Oncomine™ Breast cfDNA Research Assay v2
Variant type
Oncomine™ Gene class
Oncomine™ Variant class
Annotation criteria
Copy number amplification
Gain-of-Function
Amplification
  • SVTYPE = "CNV"
  • FILTER = "GAIN"
  • Occurs in a designated copy-gain gene
Loss-of-function truncating de novo mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Functional impact is frameshift block substitution, frameshift deletion, frameshift insertion, or nonsense
  • Occurs in a de-novo calling gene
  • Meets de-novo thresholds
Missense hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and codon position occur in predefined missense hotspot list
In-frame hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined in-frame hotspot list
Loss-of-function truncating hotspot mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined truncating hotspot list
  • Occurs in a loss-of-function gene
MNV hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript and coding syntax occur in MNV hotspot list