Oncomine™ Tumor Mutation Load Assay
Variant type
Oncomine™ Gene class
Oncomine™ Variant class
Annotation criteria
Loss-of-function truncating de novo mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Functional impact is frameshift block substitution, frameshift deletion, frameshift insertion, or nonsense
  • Occurs in a loss-of-function gene
Missense hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and codon position occur in predefined missense hotspot list
In-frame hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined in-frame hotspot list
Splice site hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript, location, and exon occur in predefined splice site hotspot list
Intronic hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript and ID occur in predefined intronic hotspot list
MNV hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript and coding syntax occur in MNV hotspot list
CEBPA‑⁠bZIP[1]
Loss-of-Function
CEBPA‑⁠bZIP
  • Positive mutation call
  • Functional impact is nonframeshift deletion, nonframeshift insertion, or nonframeshift block substitution
  • INDEL impacts codons 282–345 of CEBPA[2]
CEBPA‑⁠N‑⁠terminal [1]
Loss-of-Function
CEBPA‑⁠N‑⁠terminal
  • Positive mutation call
  • Functional impact is nonframeshift deletion, nonframeshift insertion, nonframeshift block substitution, frameshift deletion, frameshift insertion, or frameshift block substitution
  • INDEL impacts codons 1–120 of CEBPA [2]
EGFR exon 19 deletion [1]
Gain-of-Function
EGFRExon19Deletion
  • Positive mutation call
  • Functional impact is nonframeshift deletion, nonframeshift block substitution
  • Deletion impacts codons 744–761 of EGFR [2]
EGFR exon 20 insertion [1]
Gain-of-Function
EGFRExon20Insertion
  • Positive mutation call
  • Functional impact is nonframeshift insertion, nonframeshift block substitution
  • Insertion impacts codons 762–775 of EGFR or variant is in EGFR exon 20 insertion confirmed list [2]
ERBB2 exon 20 insertion [1]
Gain-of-Function
ERBB2Exon20Insertion
  • Positive mutation call
  • Functional impact is nonframeshift insertion, nonframeshift block substitution
  • Insertion impacts codons 770–783 of ERBB2 [2]
JAK2 exon 12 indel [1]
Gain-of-Function
JAK2Exon12Indel
  • Positive mutation call
  • Functional impact is nonframeshift deletion, nonframeshift insertion, or nonframeshift block substitution
  • INDEL impacts codons 536-547 of JAK2 [2]
KIT exon 11 deletion [1]
Gain-of-Function
KITExon11Deletion
  • Positive mutation call
  • Functional impact is nonframeshift deletion, or nonframeshift block substitution
  • Deletion impacts codons 550-580 of KIT [2]
  • Alternatively, mutation occurs in splice site flanking the 5' end of exon 11
KIT exon 11 insertion [1]
Gain-of-Function
KITExon11Insertion
  • Positive mutation call
  • Functional impact is nonframeshift insertion, or nonframeshift block substitution
  • Insertion impacts codons 550-580 of KIT [2]
MET exon 14 skipping [1]
Gain-of-Function
METExon14Skipping
  • Positive mutation call
  • Location is splice site in MET exon 14, is intronic >= 4bp deletion in 30 nucleotides preceding MET exon 14, or variant is in MET exon 14 skipping confirmed list
PDGFRA exon 18 deletion [1]
Gain-of-Function
PDGFRAExon18Deletion
  • Positive mutation call
  • Functional impact is nonframeshift deletion, or nonframeshift block substitution
  • Deletion impacts codons 841-847 of PDGFRA [2]
1 For more information, see Ion Torrent class-based variants.
2 In some cases the assay design may not cover the entire codon range.