Oncomine™ Myeloid MRD Assays
Variant type
Oncomine™ Gene class
Oncomine™ Variant class
Annotation criteria
Gene fusion
Gain-of-Function
Fusion
  • SVTYPE = "Fusion"
  • FILTER = "PASS"
  • Is a targeted fusion isoform
Non-targeted fusion
Gain-of-Function
NonTargetedFusion
  • SVTYPE = "Fusion"
  • FILTER = "PASS"
  • Non-Targeted field is present
  • NT_FUSION_IN_FRAME is not FALSE
  • NT_FUSION_DRIVER_INVOLVED is not FALSE
  • NT_FUSION_SECONDARY is not TRUE
RNA exon variant
Gain-of-Function
RNAExonVariant
  • SVTYPE = "RNAExonVariant" or "Fusion"
  • FILTER = "PASS"
  • Is a targeted RNA exon variant
Loss-of-function truncating de novo mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Functional impact is frameshift block substitution, frameshift deletion, frameshift insertion, or nonsense
  • Occurs in a de novo calling gene
Missense hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Functional impact is missense
  • Transcript and coding syntax occur in predefined missense hotspot list
In-frame hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined in-frame hotspot list
Gain-of-function truncating hotspot mutation
Gain-of-Function
Hotspot
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined truncating hotspot list
  • Occurs in a gain-of-function gene
Loss-of-function truncating hotspot mutation
Loss-of-Function
Truncating
  • Positive mutation call
  • Function, transcript, and coding syntax occur in predefined truncating hotspot list
  • Occurs in a loss-of-function gene
MNV hotspot mutation
Gain-of-Function
Loss-of-Function
Hotspot
  • Positive mutation call
  • Transcript and coding syntax occur in MNV hotspot list
FLT3 internal tandem duplication
Gain-of-Function
FLT3ITD
  • SVTYPE = "FLT3ITD"
  • SUBTYPE is not "Combined"