Ion Reporter™ Software web services API
Ion Reporter™ Software has a web services API that allows you to automate returns to retrieve key information from the system.
This section provides examples that demonstrate how to leverage the API.
-
These examples are used with the hosted Ion Reporter™ Software instance. If you are running a local version on your own Ion Reporter™ Server, then replace the hosted URL
with the URL root of your local instance.https://ionreporter.thermofisher.com/ -
API tokens that are included in examples are invalid and cannot be used for authentication.
-
If the JSON download process converts @ symbols to %40, do not change the text. Instead, copy the JSON files as is into the browser.