Start an analysis
|
startAnalysis |
|
|---|---|
|
URL |
/api/v1/startAnalysis |
|
Description |
Used by the IonReporterUploader plugin to start an analysis for the setId that is entered. Returns a JSON object with status and message. |
|
Method |
POST |
|
Request query parameters |
|
|
Example request |
|
|
Request headers |
Content-Type: application/x-www-form-urlencoded Authorization:API Token |
|
Returns |
|
Response fields
|
Status code |
Response message |
|---|---|
|
200 |
Successful response |
|
400 |
Bad request |
|
404 |
Not found |
|
401 |
User is not authenticated |
|
500 |
Error occurred |
Curl command
curl -X POST -k https://<server name>/api/v1/startAnalysis -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw" -d "setId=4__f535ccde-5c6a-490d-a1d2-7b81352e6830_2016-6-17_1_13_24&containerName=New_Analysis_Name"