|
PDF Report
|
FileName format is “signature_identification_<analysis_name>_report.pdf”. The PDF report consists of 4 sections:
-
Header
-
Report Name - "Mutational Signature Prediction Report"
-
Sample Name
-
Analysis Name (If a sample is re-analysed, the Analysis name will change)
-
Summary
-
Mutation Profiles
-
Signature Specific Gene Mutations
|
|
Profile of Mutational signature of Sample
|
File name format is “<analysis_name>_normalized_sample_profile.png”. The image is present in the PDF report under “Mutation Profile” section.
|
|
Profile of COSMIC signature
|
File name format is “<analysis_name>_<cosmic_signature>.png”. The image is present in the PDF report under “Mutation Profile” section. One image is generated per enriched COSMIC signature. The output will not be generated if any enriched signature is not found.
|
|
deconstructSigs Weights Pie Chart
|
File name format is “<analysis_name>_signatures_pie.png”. The image is present in the PDF report under “Summary” section. The output will not be generated if any enriched signature is not found.
|
|
Trinucleotide distribution of Variants
|
File name format is “<analysis_name>_contextFile.txt”. It contains frequencies of mutations within each of the 96 trinucleotides.
|