Preferred transcripts in default analysis workflows
The default analysis workflows that are provided in Ion Reporter™ Software include an annotations set with a preferred transcript set, the RefGene canonical transcripts. The default annotation set that includes the default RefSeq canonical transcripts is named All.
Default analysis workflows that use the All annotation set report only transcripts that are included in the RefGene canonical transcripts. By default, the following RefGene canonical transcripts are included in the All annotation set:
RefGene
Functional
Canonical
Transcript Scores
All other transcripts are filtered out and not reported.