Create annotation set without preferred transcript sets
  1. In the Workflows tab, click Presets.
  2. Click Create Preset > Annotation Set.
    1. In the Reference drop-down menu, select Gene Model as the type of annotation source.
    2. In the Choose Existing tab, click Use for either the RefSeq Gene Model or the Ensembl® Gene Model.
      Note: Only one gene model can be added to each annotation set.
  3. Enter a descriptive name for the custom annotation set.
  4. (Optional) Enter a description for the custom annotation set.
  5. Click Save.
    The new annotation source is listed in the Workflow Presets screen.