JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. annotation-only analysis workflow This predefined analysis workflow adds annotations when a VCF file is uploaded to Ion Reporter™ Software; there is no further analysis of the data, and no variants are called in Ion Reporter™ Software with this analysis workflow. ×