Restore table preference default settings
You can return the Sample table to default settings in Ion Reporter™ Software.
  1. In the Samples tab, click Overview.
  2. Click PreferencesRestore Defaults.
The table is returned to the default column settings. For the list of default columns, see Sample attributes.