Rearrange columns in the Samples table
You can change the order of sample attribute columns in your Samples table in Ion Reporter™ Software. You can use the rearranged table temporarily, or to save the column order and use it later, you can create a table preference.
  1. In the Samples tab, click Overview.
  2. Click the column heading that you want to move, then drag and drop the heading to the position you prefer.
  3. Repeat to move additional columns.
  4. Click PreferencesSave Table Preference As.
  5. In Save Table Preference, enter a name for the table preference, then click Save.
The sample table preference is saved.
To use the table preference to return to the saved view of the Samples table, or toggle between different views of the table. see Apply a preference to the Samples table.