Get VCF files
getvcf
URL
/api/v1/getvcf
Description
Gets the links to download VCF files. Returns a JSON object that contains the download information.
Method
GET
Request query parameters
  • format
    String. Optional.
    Response type, current version supports only json.
  • type
    String. Optional.
    Valid values are "sample" or "analysis".
  • name
    String. Conditional.
    If type parameter is "sample" then name is Mandatory.
  • id
    String. Optional.
    Id of an Analysis.
  • start_date
    String. Optional.
    Parameter to filter the results by date boundary. Date format should be in yyyy-MM-dd.
  • end_date
    String. Optional.
    Parameter to filter the results by date boundary. Date format should be in yyyy-MM-dd.
  • duration
    String. Optional. Parameter to filter the results by days/months.
    Valid value starts with "-" followed by numeric value and ends with "d" or "m"
  • exclude
    String. Optional. Parameter to filter the response.
    Valid values are unfilteredvariants or filteredvariants.
Example request
https://ionreporter.thermofisher.com/api/v1/getvcf?format=json&name=xyzsampletest
&start_date=2016-01-01&end_date=2016-02-01
Request headers
Content-Type:application/x-www-form-urlencoded
Authorization:API Token
Returns
[
  {
    "data_links": "http://ionreporter.thermofisher.com/api/v1/download?
      filePath=/data/IR/data/IR_Org/ion.reporter@lifetech.com/JohnSmithSample
      /JohnSmithSample_20160429014705727/JohnSmithSample_c150_2016-04-29-14-16-534.zip",
    "name": "JohnSmithSample_c150_2016-04-29-14-16-534",
    "id": "ff808181545d90790154613336be0008"
  }
]
Response fields
Status code
Response message
200
Successful response
400
Bad request
404
Not found
401
User is not authenticated
500
Error occurred
Curl command
curl -v -k -X GET "https://<server name>/api/v1/getvcf?format=json&name=BRCA_Plus_RR_jul29&start_date=2016-07-25&end_date=2016-07-31" -H "Content-Type:application/x-www-form-urlencoded" -H "Authorization:NDliYmU0OWE4MzA3YmE2MWVmMjhlZTlhOTEyMmE5ODM0NzU3NjQ4OGM5NzVjODU3YjdlMWM4Mjk3OGYzODE4Yw"