Create a MyVariants and Variant Classification filter chain
You can create a custom filter chain to narrow the analysis results to include only the variants that are flagged and classified by members of your Ion Reporter™ Software organization. This example shows how to set up a filter chain for MyVariants that are flagged as Important and classified as Deleterious in Ion Reporter™ Software.
IMPORTANT! If using a single filter with a NOT operator, do not use a parenthesis. If parenthesis is used, the filter chain can be saved but an error occurs when applying the filter chain in analyses results. For example, use NOT Filtered Coverage instead of NOT (Filtered Coverage).
In the Workflows tab, click Presets.
Click Create Preset > Filter Chain.
In the Create Filter Chain dialog box, enter a name and optional description.
In the Choose Filter list, select MyVariants.
Select Important.
Click Set.
In the Choose Filter list, select Variant Classification.