Copy analyses to storage on Connect
You can copy VCF files from successful Ion Reporter™ Software analyses to storage on Connect to store and share analyses with other users. You can also upload analyses to use with other software applications that are available on Connect, such as Oncomine™ Reporter Software. For more information, see Increase storage space on Connect.
- In the Analyses tab, click Overview.
- Select the row that contains the successful analysis that you want to copy to storage on Connect.
- In the Details pane, click .
-
In the Export to Data Manager dialog box, click Confirm.
A message at the top of the Analyses screen indicates that the files have been copied to storage on Connect.
- To view the files, sign in to Connect, then click .
×