Reanalyze aborted analysis results
You can reanalyze results from any failed or aborted analysis that uses an analysis workflow that is supported in the version of Ion Reporter™ Software that you use. To view details about an analysis workflow, see View analysis workflow details.
  1. In the Analyses tab, select an analysis of interest that has a status of Aborted.
  2. Click ActionsReanalyze.