The Analyses table includes details that describe the samples, analysis workflows, and software versions that were used to create the analyses in Ion Reporter™ Software.
The information that is shown in the table depends on which analysis workflow and annotation set are applied when the analysis is launched. You can customize the table to reflect the information that is most relevant to your research. To customize the table, create a table preference, or drag and drop the columns to change the order in which they appear.