p-value
Probability value. A statistical method for the detection of variant calls from next-generation sequencers.
In Ion Reporter™ Software, p-values in Analysis Results in the column named p-value are rounded to 5 decimal places (between 0.00001-0.99999) when shown in the software screen. Very small p-values that are less than 0.00001 are rounded to 0.00001 by default when shown in the software screen. Very large p-values that are greater than 0.99999 are rounded to 0.99999 by default when shown in the software screen.