JavaScript must be enabled in order to use this site.Please enable JavaScript in your browser and refresh the page. Genetic Category Type and MAF filter If a variant is not a minor allele, MAF filter will not be effective, because MAF filter only applies if the variant is a minor allele. Users may have a threshold setting for MAF filter which will not be effective if variant is not a minor allele. Related Topics Parent topic: Genetic Category Type variants ×