Filter impact on the variants displayed in
heat maps
If you save your filter chain on the Analysis Results page, then the heat maps and other multiple analysis visualizations show only filtered in variants.
If you do not save your filter on the Analysis Results page, then the heat maps and other multiple analysis visualizations show only the entire result sets (nothing is filtered out).