Adjust IRGV BAM tracks
You can adjust the BAM read coverage track in an Ion Reporter™ Genomic Viewer (IRGV) visualization to make it easier to view the data. When you adjust the BAM read coverage track, it changes the amount of data that is shown in the plot. For example, you can see more BAM tracks when you use the Squish setting.
For best results, use the Expanded option to click and view hover help with details about variants and base calls that are visualized in each read coverage track.
Option |
Description |
---|---|
Select this option to view the read coverage track with the maximum visible height for each BAM track. |
|
Select this option to view reads in the coverage track in a single line that has minimal height for each BAM track. |
|
Select this option to view no BAM tracks and view only the coverage density at the top of the BAM track. |